Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288987.1 | NP_001275916.1 | 4, 5 | 438 | ||
NM_014581.3 | NP_055396.1 | 4, 5 | 438 | |||
NR_110242.1 | 5, 6 | 544 | ||||
NR_110243.1 | 3, 4 | 304 | ||||
XM_006717086.3 | XP_006717149.1 | 4, 5 | 506 | |||
XM_017014654.1 | XP_016870143.1 | 5, 6 | 674 | |||
XM_017014655.1 | XP_016870144.1 | 6, 7 | 885 | |||
GenBank mRNA | AJ251026.1 | - | 4, 5 | 429 | ||
AJ251027.1 | - | 5, 6 | 535 | |||
AJ251028.1 | - | 3, 4 | 295 | |||
AY358981.1 | - | 4, 5 | 431 | |||
BC096717.1 | - | 4, 5 | 413 | 40003080 | ||
BC098125.1 | - | 4, 5 | 413 | 40003075 | ||
BC098151.1 | - | 4, 5 | 413 | 40003079 | ||
BC098340.1 | - | 4, 5 | 413 | 40003073 | ||
BX101545.1 | - | 2, 3 | 159 | |||
HY046406.1 | - | 4, 5 | 438 |