Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020890.2 | NP_065941.2 | 9, 10 | 1378 | ||
XM_006713716.3 | XP_006713779.1 | 9, 10 | 1162 | |||
XM_006713717.2 | XP_006713780.1 | 10, 11 | 1274 | |||
XM_011513056.1 | XP_011511358.1 | 9, 10 | 1122 | |||
XM_011513057.2 | XP_011511359.1 | 10, 11 | 1243 | |||
GenBank mRNA | AB040957.1 | - | 9, 10 | 1105 | ||
AF334474.1 | - | 9, 10 | 1177 | |||
AK056933.1 | - | 9, 10 | 1187 | |||
AK308315.1 | - | 9, 10 | 1124 | |||
AK310446.1 | - | 9, 10 | 1382 | |||
AL713689.1 | - | 9, 10 | 1173 | |||
BC063433.1 | - | 9, 10 | 1169 | |||
BC082961.1 | - | 9, 10 | 1131 | |||
BC130564.1 | - | 9, 10 | 1145 | 40146595 | ||
BC136371.1 | - | 9, 10 | 1145 | 9020358 |