Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBNL1 SIRNA Assays ›
Gene Symbol:
MBNL1
Gene Name:
muscleblind like splicing regulator 1
Gene Aliases:
EXP, MBNL
Chromosome Location:
Chr.3: 152243656 - 152465780 on Build GRCh38
UniGene ID:
Hs.201858
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
29028
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBNL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001314057.1
NP_001300986.1
12
2603
NM_021038.4
NP_066368.2
10
3334
NM_207292.2
NP_997175.1
9
3298
NM_207293.1
NP_997176.1
9
2727
NM_207294.1
NP_997177.1
7
2469
NM_207295.1
NP_997178.1
8
2505
NM_207296.1
NP_997179.1
8
2583
NM_207297.1
NP_997180.1
8
2614
XM_005247457.4
XP_005247514.1
12
2956
XM_005247458.4
XP_005247515.1
12
4210
XM_005247460.4
XP_005247517.1
12
2919
XM_005247461.4
XP_005247518.1
11
2920
XM_005247462.4
XP_005247519.1
11
2902
XM_005247463.4
XP_005247520.1
10
2866
XM_005247464.4
XP_005247521.1
11
3014
XM_005247465.4
XP_005247522.1
11
2861
XM_005247466.4
XP_005247523.1
10
2955
XM_005247467.4
XP_005247524.1
10
2825
XM_005247468.4
XP_005247525.1
9
2771
XM_005247471.4
XP_005247528.1
9
2837
XM_005247474.3
XP_005247531.1
10
1874
XM_005247475.3
XP_005247532.1
11
2197
XM_005247476.4
XP_005247533.1
9
1910
XM_005247477.4
XP_005247534.1
10
1963
XM_006713639.1
XP_006713702.1
12
2938
XM_011512843.2
XP_011511145.1
11
2752
XM_011512845.2
XP_011511147.1
10
2698
XM_011512846.2
XP_011511148.1
9
2662
XM_011512847.2
XP_011511149.1
9
2661
XM_011512848.2
XP_011511150.1
12
2560
XM_011512849.2
XP_011511151.1
8
2569
XM_017006423.1
XP_016861912.1
11
3207
XM_017006424.1
XP_016861913.1
11
3034
XM_017006425.1
XP_016861914.1
10
3000
XM_017006426.1
XP_016861915.1
10
2998
XM_017006427.1
XP_016861916.1
10
3127
XM_017006428.1
XP_016861917.1
10
2791
XM_017006429.1
XP_016861918.1
10
2874
XM_017006430.1
XP_016861919.1
9
2902
XM_017006431.1
XP_016861920.1
9
2979
XM_017006432.1
XP_016861921.1
10
2896
XM_017006433.1
XP_016861922.1
9
2860
XM_017006434.1
XP_016861923.1
9
2866
XM_017006435.1
XP_016861924.1
11
2004
XM_017006436.1
XP_016861925.1
9
3968
XM_017006437.1
XP_016861926.1
8
2987
XM_017006438.1
XP_016861927.1
9
2623
XM_017006439.1
XP_016861928.1
10
2143
XM_017006440.1
XP_016861929.1
9
1877
XM_017006441.1
XP_016861930.1
8
1874
XM_017006442.1
XP_016861931.1
8
1977
XM_017006443.1
XP_016861932.1
7
1890
GenBank mRNA
AB007888.1
-
9
3298
AF255334.1
-
7
2191
AK297133.1
-
6
1389
AK307420.1
-
7
1429
BC043493.1
-
10
2839
6044671
BC050535.1
-
12
2124
6042311
BX537491.1
-
2
2407
CR749454.1
-
4
962
Y13829.1
-
9
2449
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
mRNA processing
nervous system development
RNA splicing
embryonic limb morphogenesis
regulation of RNA splicing
myoblast differentiation
regulatory region RNA binding
RNA binding
double-stranded RNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-rzq2w:80/10.239.163.191:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0