Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANBAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003897.1 | NP_001003897.1 | 3 | 304 | ||
NM_022077.3 | NP_071360.1 | 4 | 421 | |||
XM_017028006.1 | XP_016883495.1 | 4 | 1479 | |||
XM_017028007.1 | XP_016883496.1 | 3 | 1362 | |||
GenBank mRNA | AK026708.1 | - | 4 | 1021 | ||
AK292971.1 | - | 4 | 422 | |||
BC014672.1 | - | 3 | 282 | 3535633 | ||
BC016822.1 | - | 5 | 645 | 4102182 | ||
BI754866.1 | - | 3 | 304 | |||
BQ051237.1 | - | 423 |