Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCUR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031713.3 | NP_001026883.1 | 2 | 597 | ||
XM_011514802.1 | XP_011513104.1 | 2 | 600 | |||
GenBank mRNA | AK024611.1 | - | 2 | 606 | ||
BC008234.1 | - | 2 | 352 | |||
BC016850.1 | - | 2 | 571 | 3886144 | ||
BP382285.1 | - | 2 | 606 |