Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543544.1 | XP_011541846.1 | 2 | 591 | ||
GenBank mRNA | AB109031.1 | - | 2 | 558 | ||
AB163917.1 | - | 2 | 558 | |||
AF191545.1 | - | 1 | 530 | |||
AK074281.1 | - | 2 | 635 | |||
AK312864.1 | - | 2 | 571 | |||
AY028805.1 | - | 1 | 488 | |||
BC017927.2 | - | 1 | 531 | |||
BC065240.1 | - | 2 | 552 | 6166289 |