Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POPDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022361.4 | NP_071756.2 | 4 | 1156 | ||
NR_024539.1 | 4 | 697 | ||||
XM_011536067.2 | XP_011534369.1 | 4 | 1316 | |||
XM_017011194.1 | XP_016866683.1 | 4 | 2599 | |||
XM_017011195.1 | XP_016866684.1 | 4 | 2059 | |||
GenBank mRNA | AF204171.1 | - | 3 | 901 | ||
AK225989.1 | - | 4 | 697 | |||
AK314101.1 | - | 4 | 1156 | |||
BC022323.2 | - | 3 | 1006 | 4293961 |