Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032396.2 | NP_001027568.1 | 2 | 2188 | ||
NM_022368.4 | NP_071763.2 | 3 | 1950 | |||
NM_145119.3 | NP_660095.1 | 2 | 2514 | |||
XM_005262292.1 | XP_005262349.1 | 2 | 2176 | |||
XM_011531011.2 | XP_011529313.1 | 2 | 2321 | |||
GenBank mRNA | AF262024.1 | - | 1 | 2443 | ||
AF264620.1 | - | 3 | 1865 | |||
AK021892.1 | - | 1 | 1293 | |||
AK096611.1 | - | 2 | 2513 | |||
AK290543.1 | - | 2 | 2414 | |||
AK294632.1 | - | 3 | 2027 | |||
BC039149.1 | - | 1 | 1520 | |||
BC048323.1 | - | 1 | 1360 | |||
BC058327.1 | - | 1 | 1500 | |||
BC075803.1 | - | 2 | 2421 | 3913310 |