Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTBP2 SIRNA Assays ›
Gene Symbol:
CTBP2
Gene Name:
C-terminal binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.10: 124987849 - 125161061 on Build GRCh38
UniGene ID:
Hs.501345
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
30207
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083914.2
NP_001077383.1
4, 5
646
NM_001290214.2
NP_001277143.1
4, 5
631
NM_001290215.2
NP_001277144.1
4, 5
895
NM_001321012.1
NP_001307941.1
4, 5
590
NM_001321013.1
NP_001307942.1
3, 4
791
NM_001321014.1
NP_001307943.1
4, 5
892
NM_001329.3
NP_001320.1
4, 5
634
NM_022802.2
NP_073713.2
2, 3
1951
XM_005269561.2
XP_005269618.1
5, 6
817
XM_005269564.2
XP_005269621.1
5, 6
1025
XM_005269567.2
XP_005269624.1
5, 6
1024
XM_005269568.4
XP_005269625.1
5, 6
752
XM_005269569.2
XP_005269626.1
4, 5
713
XM_005269571.2
XP_005269628.1
3, 4
472
XM_005269572.3
XP_005269629.1
3, 4
520
XM_006717642.2
XP_006717705.1
6, 7
946
XM_011539349.2
XP_011537651.1
2, 3
728
XM_011539351.1
XP_011537653.1
7, 8
1047
XM_011539353.1
XP_011537655.1
6, 7
918
XM_011539354.1
XP_011537656.1
5, 6
842
XM_011539355.1
XP_011537657.1
4, 5
920
XM_011539357.1
XP_011537659.1
4, 5
658
XM_011539358.1
XP_011537660.1
3, 4
736
XM_017015756.1
XP_016871245.1
5, 6
688
XM_017015757.1
XP_016871246.1
4, 5
2414
GenBank mRNA
AF016507.1
-
4, 5
551
AF222711.1
-
2, 3
1968
AK290390.1
-
4, 5
495
AL833398.1
-
4, 5
606
BC002486.2
-
4, 5
607
BC037900.2
-
2, 3
615
5276086
BC047018.1
-
4, 5
611
BC052276.1
-
4, 5
588
BC072020.1
-
4, 5
568
BM800106.1
-
2, 3
306
BT007012.1
-
2, 3
206
CV029885.1
-
2, 3
206
CX165167.1
-
4, 5
460
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of cell proliferation
viral genome replication
positive regulation of chromatin binding
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of retinoic acid receptor signaling pathway
white fat cell differentiation
oxidation-reduction process
chromatin binding
transcription coactivator activity
transcription corepressor activity
protein binding
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
protein homodimerization activity
retinoic acid receptor binding
NAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9zxgn:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0