Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324475.1 | NP_001311404.1 | 19, 20 | 2146 | ||
NM_023015.4 | NP_075391.3 | 18, 19 | 2487 | |||
XM_005245459.2 | XP_005245516.1 | 18, 19 | 2504 | |||
XM_005245461.2 | XP_005245518.1 | 12, 13 | 1177 | |||
XM_006711490.3 | XP_006711553.1 | 19, 20 | 1957 | |||
XM_011509906.2 | XP_011508208.1 | 18, 19 | 1869 | |||
XM_017002132.1 | XP_016857621.1 | 8, 9 | 745 | |||
GenBank mRNA | AK025572.1 | - | 4, 5 | 308 | ||
AK074979.1 | - | 11, 12 | 1185 | |||
AK225294.1 | - | 4, 5 | 308 | |||
AK290025.1 | - | 19, 20 | 2140 | |||
AK298746.1 | - | 6, 7 | 637 | |||
AK304874.1 | - | 12, 13 | 1576 | |||
AL080172.1 | - | 13, 14 | 1386 | |||
AL832133.1 | - | 17, 18 | 3522 | |||
BC025254.1 | - | 2, 3 | 147 | |||
BC054513.1 | - | 5, 6 | 523 | |||
BC073985.1 | - | 2, 3 | 159 | |||
BC098431.1 | - | 4, 5 | 347 | |||
BC105092.1 | - | 18, 19 | 1950 | 8144095 | ||
BC105094.1 | - | 18, 19 | 1950 | 8144097 | ||
BC116458.1 | - | 18, 19 | 2482 | 5752962 | ||
BX640786.1 | - | 18, 19 | 2473 | |||
BX640950.1 | - | 18, 19 | 2488 | |||
CR749212.1 | - | 16, 17 | 4014 | |||
CR749376.1 | - | 17, 18 | 4139 |