Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006342.2 | NP_006333.1 | 13 | 2245 | ||
XM_005247929.2 | XP_005247986.1 | 13 | 2255 | |||
XM_005247930.2 | XP_005247987.1 | 12 | 1284 | |||
XM_017007653.1 | XP_016863142.1 | 13 | 2297 | |||
GenBank mRNA | AF093543.1 | - | 13 | 2198 | ||
AF479620.1 | - | 12 | 1111 | |||
AJ243997.1 | - | 13 | 2190 | |||
BC106071.1 | - | 13 | 2206 | 6148176 | ||
BC111771.1 | - | 13 | 2158 | 40034847 |