Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317929.1 | NP_001304858.1 | 10 | 2236 | ||
NM_024330.1 | NP_077306.1 | 10 | 2341 | |||
GenBank mRNA | AJ577572.1 | - | 10 | 2634 | ||
AK027499.1 | - | 10 | 2245 | |||
AK074134.1 | - | 2 | 4436 | |||
AK075377.1 | - | 10 | 2334 | |||
AK123997.1 | - | 11 | 1762 | |||
AK125102.1 | - | 10 | 3278 | |||
AY358409.1 | - | 10 | 2329 | |||
BC003041.1 | - | 10 | 2341 | 2822875 | ||
BC003654.2 | - | 9 | 1285 | |||
BC009916.2 | - | 10 | 2341 | 2822875 | ||
BC029792.1 | - | 10 | 2209 | 5195359 |