Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTDC1 SIRNA Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
Hmat-Xa, mat-Xa
Chromosome Location:
Chr.2: 143945787 - 144333092 on Build GRCh38
UniGene ID:
Hs.44780
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31369
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006636.3
NP_001006637.1
7, 8
1159
NM_001164629.3
NP_001158101.1
7, 8
1144
NM_001284233.1
NP_001271162.1
5, 6
968
NM_001284234.1
NP_001271163.1
5, 6
859
NM_001284238.1
NP_001271167.1
9, 10
1358
XM_005263774.2
XP_005263831.1
8, 9
1243
XM_005263782.2
XP_005263839.1
8, 9
1259
XM_005263784.2
XP_005263841.1
6, 7
1118
XM_011511843.2
XP_011510145.1
8, 9
1292
XM_011511855.2
XP_011510157.1
6, 7
1236
XM_011511856.2
XP_011510158.1
6, 7
1018
XM_017004917.1
XP_016860406.1
12, 13
1914
XM_017004918.1
XP_016860407.1
10, 11
1506
XM_017004919.1
XP_016860408.1
8, 9
1392
XM_017004920.1
XP_016860409.1
10, 11
4548
XM_017004921.1
XP_016860410.1
9, 10
1341
XM_017004922.1
XP_016860411.1
9, 10
1518
XM_017004923.1
XP_016860412.1
8, 9
1306
XM_017004924.1
XP_016860413.1
10, 11
1502
XM_017004925.1
XP_016860414.1
7, 8
1164
XM_017004926.1
XP_016860415.1
5, 6
1095
XM_017004927.1
XP_016860416.1
7, 8
1126
XM_017004928.1
XP_016860417.1
9, 10
1359
XM_017004929.1
XP_016860418.1
7, 8
1096
XM_017004930.1
XP_016860419.1
7, 8
1747
XM_017004931.1
XP_016860420.1
7, 8
1160
XM_017004932.1
XP_016860421.1
7, 8
1130
XM_017004933.1
XP_016860422.1
7, 8
1112
XM_017004934.1
XP_016860423.1
8, 9
4319
XM_017004935.1
XP_016860424.1
9, 10
2189
XM_017004936.1
XP_016860425.1
8, 9
1366
XM_017004937.1
XP_016860426.1
6, 7
1247
XM_017004939.1
XP_016860428.1
8, 9
1295
XM_017004940.1
XP_016860429.1
5, 6
1096
XM_017004942.1
XP_016860431.1
7, 8
1142
XM_017004943.1
XP_016860432.1
7, 8
1808
GenBank mRNA
AB201269.1
-
4, 5
880
AB597184.1
-
4, 5
880
AK021815.1
-
1, 2
335
AK091684.1
-
5, 6
972
AK291357.1
-
7, 8
1074
AY281366.1
-
7, 8
1159
BX647436.1
-
5, 6
841
BX647773.1
-
9, 10
1297
Back To Top
More Information
Gene Ontology Categories:
Process(es)
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-bn9m5:80/10.239.167.51:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0