Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YRDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024640.3 | NP_078916.3 | 5 | 1455 | ||
GenBank mRNA | AF250397.1 | - | 5 | 1274 | ||
AK027129.1 | - | 4 | 1059 | |||
AK129676.1 | - | 1 | 2281 | |||
AY172561.1 | - | 5 | 1447 | |||
AY286019.1 | - | 5 | 1435 | |||
BC067857.1 | - | 5 | 1440 | |||
BC080186.1 | - | 5 | 1457 | 6147134 |