Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024734.3 | NP_079010.2 | 2 | 231 | ||
XM_011537158.1 | XP_011535460.1 | 2 | 227 | |||
XM_011537159.2 | XP_011535461.1 | 2 | 227 | |||
XM_017021646.1 | XP_016877135.1 | 2 | 397 | |||
XM_017021647.1 | XP_016877136.1 | 2 | 227 | |||
GenBank mRNA | AB047979.1 | - | 2 | 188 | ||
AK025226.1 | - | 2 | 174 | |||
AK314315.1 | - | 2 | 229 | |||
AL117526.1 | - | 2 | 83 | |||
BC068482.1 | - | 2 | 180 | 4375469 | ||
CB155696.1 | - | 2 | 231 |