Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM192A SIRNA Assays ›
Gene Symbol:
FAM192A
Gene Name:
family with sequence similarity 192 member A
Gene Aliases:
C16orf94, CDA018, CDA10, NIP30
Chromosome Location:
Chr.16: 57152466 - 57186242 on Build GRCh38
UniGene ID:
Hs.396740
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31893
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM192A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024946.2
NP_079222.1
2
347
XM_005256156.4
XP_005256213.1
2
315
XM_005256158.3
XP_005256215.1
2
1402
XM_005256160.2
XP_005256217.1
2
319
XM_005256163.2
XP_005256220.1
3
456
XM_005256164.3
XP_005256221.1
3
401
XM_005256165.2
XP_005256222.1
2
376
XM_006721275.3
XP_006721338.2
2
1085
XM_011523343.2
XP_011521645.1
2
193
XM_017023681.1
XP_016879170.1
2
411
XM_017023682.1
XP_016879171.1
3
1520
XM_017023683.1
XP_016879172.1
3
1471
XM_017023684.1
XP_016879173.1
3
686
XM_017023685.1
XP_016879174.1
3
1435
XM_017023686.1
XP_016879175.1
4
1505
XM_017023687.1
XP_016879176.1
4
1480
XM_017023688.1
XP_016879177.1
4
1423
XM_017023689.1
XP_016879178.1
4
804
XM_017023690.1
XP_016879179.1
4
755
XM_017023691.1
XP_016879180.1
4
808
XM_017023692.1
XP_016879181.1
3
592
XM_017023693.1
XP_016879182.1
3
495
XM_017023694.1
XP_016879183.1
4
598
XM_017023695.1
XP_016879184.1
4
710
XM_017023696.1
XP_016879185.1
3
1093
XM_017023697.1
XP_016879186.1
3
478
XM_017023698.1
XP_016879187.1
3
1201
XM_017023699.1
XP_016879188.1
3
729
XM_017023700.1
XP_016879189.1
3
198
XM_017023701.1
XP_016879190.1
3
664
XM_017023702.1
XP_016879191.1
3
384
XM_017023703.1
XP_016879192.1
2
124
XM_017023704.1
XP_016879193.1
2
122
GenBank mRNA
AF212249.1
-
2
300
AF271785.1
-
2
300
AF356585.1
-
1
98
AK023291.1
-
2
247
AK025452.1
-
2
322
AL834421.1
-
2
159
BC020536.1
-
2
315
3849620
BC063409.1
-
2
255
BC071952.1
-
1
116
6305656
BI545617.1
-
2
352
DB101263.1
-
2
322
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001014014, XM_006255111, XM_006255114, XM_006255110, BC079145.This siRNA also targets the following transcripts in Mouse: NM_028221, AK050704, BC016088, BC023158, AK033988, and others.
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0