Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001268284.2 | NP_001255213.1 | 4 | 3236 | ||
NM_001268285.2 | NP_001255214.1 | 2 | 2950 | |||
NM_025061.5 | NP_079337.2 | 3 | 3093 | |||
XM_011528319.2 | XP_011526621.1 | 3 | 3821 | |||
GenBank mRNA | AK027073.1 | - | 1 | 2042 | ||
AK091134.1 | - | 3 | 3090 | |||
AL834474.1 | - | 1 | 2472 | |||
BC022216.1 | - | 1 | 2378 | |||
BC070089.1 | - | 3 | 3094 | 30346745 | ||
BC108252.1 | - | 1 | 2204 | |||
BX538180.1 | - | 3 | 3084 |