Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FHOD3 SIRNA Assays ›
Gene Symbol:
FHOD3
Gene Name:
formin homology 2 domain containing 3
Gene Aliases:
FHOS2, Formactin2
Chromosome Location:
Chr.18: 36297696 - 36780220 on Build GRCh38
UniGene ID:
Hs.436636
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
32380
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FHOD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281739.2
NP_001268668.1
17, 18
3331
NM_001281740.2
NP_001268669.1
21, 22
3907
NM_025135.4
NP_079411.2
18, 19
3382
XM_005258349.1
XP_005258406.1
19, 20
3700
XM_005258354.1
XP_005258411.1
17, 18
3250
XM_005258355.1
XP_005258412.1
17, 18
3250
XM_011526189.1
XP_011524491.1
22, 23
3949
XM_011526190.1
XP_011524492.1
22, 23
3949
XM_011526192.1
XP_011524494.1
21, 22
3874
XM_011526193.2
XP_011524495.1
22, 23
3949
XM_011526195.1
XP_011524497.1
21, 22
3652
XM_011526196.1
XP_011524498.1
19, 20
3424
XM_011526197.1
XP_011524499.1
21, 22
3874
XM_017026006.1
XP_016881495.1
22, 23
3949
XM_017026007.1
XP_016881496.1
20, 21
3587
XM_017026008.1
XP_016881497.1
22, 23
3949
XM_017026009.1
XP_016881498.1
18, 19
3364
XM_017026010.1
XP_016881499.1
20, 21
3593
GenBank mRNA
AB051482.1
-
11, 12
2523
AB084087.1
-
17, 18
3191
AK025950.1
-
3, 4
477
AK091899.1
-
10, 11
1433
AL834480.1
-
1, 2
169
BC031518.1
-
14, 15
2834
BC050670.1
-
2, 3
253
BC081563.1
-
3, 4
509
HM191478.1
-
21, 22
3767
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of actin filament polymerization
sarcomere organization
actin filament network formation
cardiac myofibril assembly
actin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0