Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267803.1 | NP_001254732.1 | 4 | 280 | ||
NM_001267804.1 | NP_001254733.1 | 4 | 271 | |||
NM_001267805.1 | NP_001254734.1 | 4 | 267 | |||
NM_001267806.1 | NP_001254735.1 | 3 | 154 | |||
NM_025155.2 | NP_079431.1 | 3 | 211 | |||
XM_005274309.1 | XP_005274366.1 | 2 | 111 | |||
XM_017018357.1 | XP_016873846.1 | 4 | 240 | |||
XM_017018358.1 | XP_016873847.1 | 4 | 245 | |||
GenBank mRNA | AF087895.1 | - | 4 | 205 | ||
AK021910.1 | - | 3 | 192 | |||
AK222501.1 | - | 3 | 192 | |||
AK225520.1 | - | 3 | 179 | |||
AK298258.1 | - | 3 | 154 | |||
AK316394.1 | - | 3 | 425 | |||
BC021541.1 | - | 3 | 175 | 3942680 | ||
BC028628.1 | - | 4 | 245 | 5263875 | ||
CD243380.1 | - | 4 | 257 | |||
DC354686.1 | - | 3 | 213 |