Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSGA10 SIRNA Assays ›
Gene Symbol:
TSGA10
Gene Name:
testis specific 10
Gene Aliases:
CEP4L, CT79
Chromosome Location:
Chr.2: 98997261 - 99154962 on Build GRCh38
UniGene ID:
Hs.120267
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
32537
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSGA10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_025244.2
NP_079520.1
7
825
NM_182911.3
NP_878915.2
6
804
XM_005264023.1
XP_005264080.1
6
779
XM_005264024.2
XP_005264081.1
8
944
XM_005264025.1
XP_005264082.1
7
915
XM_005264026.1
XP_005264083.1
8
900
XM_005264027.1
XP_005264084.1
6
735
XM_005264029.1
XP_005264086.1
6
724
XM_006712781.2
XP_006712844.1
9
978
XM_011511934.1
XP_011510236.1
6
814
XM_011511935.2
XP_011510237.1
5
588
XM_011511937.1
XP_011510239.1
6
785
XM_011511939.1
XP_011510241.1
9
1073
XM_011511940.2
XP_011510242.1
8
1044
XM_011511941.1
XP_011510243.1
9
1117
XM_011511942.1
XP_011510244.1
9
1107
XM_011511943.2
XP_011510245.1
5
555
XM_017005035.1
XP_016860524.1
6
805
XM_017005036.1
XP_016860525.1
6
814
XM_017005037.1
XP_016860526.1
6
814
XM_017005038.1
XP_016860527.1
6
814
XM_017005039.1
XP_016860528.1
9
967
XM_017005040.1
XP_016860529.1
8
1068
XM_017005041.1
XP_016860530.1
6
926
XM_017005042.1
XP_016860531.1
8
956
XM_017005043.1
XP_016860532.1
6
789
XM_017005044.1
XP_016860533.1
8
932
XM_017005045.1
XP_016860534.1
9
3119
XM_017005046.1
XP_016860535.1
8
934
XM_017005047.1
XP_016860536.1
5
842
XM_017005048.1
XP_016860537.1
5
638
GenBank mRNA
AF254756.1
-
5
800
AK304328.1
-
6
724
AK313354.1
-
5
561
AY014284.1
-
5
653
BC028366.1
-
6
805
4830604
BX647157.1
-
7
1018
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
spermatogenesis
cell projection assembly
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0