Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300927.1 | NP_001287856.1 | 4 | 6299 | ||
NM_001308366.1 | NP_001295295.1 | 5 | 6492 | |||
NM_017712.3 | NP_060182.1 | 5 | 6532 | |||
XM_006722783.3 | XP_006722846.1 | 5 | 6432 | |||
XM_011528115.1 | XP_011526417.1 | 4 | 6457 | |||
XM_017026913.1 | XP_016882402.1 | 4 | 6382 | |||
GenBank mRNA | AK025948.1 | - | 1 | 1070 | ||
BC004942.1 | - | 1 | 443 |