Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033678.3 | NP_001028850.2 | 8 | 968 | ||
NM_001160389.1 | NP_001153861.1 | 8 | 974 | |||
NM_001160390.1 | NP_001153862.1 | 8 | 956 | |||
NM_001160392.1 | NP_001153864.1 | 7 | 857 | |||
NM_001160393.1 | NP_001153865.1 | 7 | 766 | |||
NM_031472.3 | NP_113660.1 | 7 | 884 | |||
XM_005274345.4 | XP_005274402.1 | 8 | 891 | |||
XM_005274346.4 | XP_005274403.1 | 8 | 850 | |||
XM_005274347.4 | XP_005274404.1 | 7 | 767 | |||
XM_006718706.3 | XP_006718769.1 | 8 | 853 | |||
XM_006718707.3 | XP_006718770.1 | 8 | 800 | |||
XM_017018400.1 | XP_016873889.1 | 7 | 767 | |||
XM_017018401.1 | XP_016873890.1 | 6 | 728 | |||
GenBank mRNA | AK026364.1 | - | 6 | 1678 | ||
AY211494.1 | - | 8 | 924 | |||
BC005133.2 | - | 7 | 780 | 3836683 | ||
BQ672032.1 | - | 6 | 671 |