Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031942.4 | NP_114148.3 | 10 | 1659 | ||
NM_145810.2 | NP_665809.1 | 9 | 1422 | |||
XM_011511957.2 | XP_011510259.1 | 10 | 2200 | |||
GenBank mRNA | AK027628.1 | - | 10 | 1631 | ||
AK027642.1 | - | 9 | 1403 | |||
AK075134.1 | - | 8 | 2285 | |||
AK223524.1 | - | 9 | 1427 | |||
AK297097.1 | - | 8 | 1257 | |||
AL833728.1 | - | 5 | 951 | |||
AL834186.1 | - | 7 | 2113 | |||
AY029179.1 | - | 9 | 1435 | |||
BC015124.1 | - | 5 | 839 | |||
BC027966.1 | - | 9 | 1388 | 5227112 |