Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301163.1 | NP_001288092.1 | 5 | 536 | ||
NM_032842.3 | NP_116231.2 | 5 | 536 | |||
XM_005250655.1 | XP_005250712.1 | 5 | 826 | |||
XM_011516646.2 | XP_011514948.1 | 5 | 676 | |||
GenBank mRNA | AK027678.1 | - | 5 | 441 | ||
AK027709.1 | - | 5 | 446 | |||
AK074732.1 | - | 5 | 469 | |||
BC045618.1 | - | 5 | 444 | 5294911 | ||
BC057784.1 | - | 5 | 441 | 5313685 | ||
BU177118.1 | - | 5 | 481 | |||
DB021198.1 | - | 5 | 536 |