Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318834.1 | NP_001305763.1 | 16 | 2496 | ||
NM_032933.5 | NP_116322.3 | 15 | 2381 | |||
NR_134875.1 | 14 | 2332 | ||||
NR_134876.1 | 11 | 2252 | ||||
NR_134877.1 | 14 | 2335 | ||||
XM_011526233.2 | XP_011524535.1 | 16 | 2473 | |||
XM_017026045.1 | XP_016881534.1 | 14 | 2295 | |||
XM_017026049.1 | XP_016881538.1 | 15 | 2377 | |||
XM_017026050.1 | XP_016881539.1 | 15 | 2399 | |||
XM_017026051.1 | XP_016881540.1 | 15 | 2301 | |||
GenBank mRNA | AK123623.1 | - | 11 | 2242 | ||
AK126247.1 | - | 14 | 2324 | |||
BC006280.2 | - | 3 | 1604 | |||
BC054507.1 | - | 9 | 1881 | |||
BC082984.1 | - | 3 | 1730 |