Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033087.3 | NP_149078.1 | 2 | 567 | ||
NR_024532.1 | 3 | 726 | ||||
GenBank mRNA | AB161356.1 | - | 2 | 524 | ||
AK027417.1 | - | 2 | 519 | |||
AK027814.1 | - | 1 | 1319 | |||
AK074704.1 | - | 2 | 519 | |||
AK074988.1 | - | 3 | 704 | |||
AK075172.1 | - | 2 | 543 | |||
AY358697.1 | - | 2 | 311 | |||
BC017876.1 | - | 2 | 517 | 4698763 | ||
DB019023.1 | - | 2 | 569 |