Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053043.2 | NP_444271.2 | 13 | 2638 | ||
XM_005249531.1 | XP_005249588.1 | 14 | 2803 | |||
XM_005249532.1 | XP_005249589.1 | 14 | 2803 | |||
XM_005249533.1 | XP_005249590.1 | 14 | 2773 | |||
XM_005249534.1 | XP_005249591.1 | 13 | 2683 | |||
XM_005249535.1 | XP_005249592.1 | 13 | 2653 | |||
XM_011515840.1 | XP_011514142.1 | 14 | 2803 | |||
GenBank mRNA | AK301179.1 | - | 8 | 1585 | ||
AK307515.1 | - | 13 | 2468 | |||
AL137724.1 | - | 11 | 3533 |