Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYATL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220494.2 | NP_001207423.1 | 6, 7 | 733 | ||
NM_001220496.2 | NP_001207425.1 | 5, 6 | 561 | |||
NM_080661.4 | NP_542392.2 | 5, 6 | 464 | |||
XM_011545354.2 | XP_011543656.1 | 5, 6 | 580 | |||
XM_011545355.2 | XP_011543657.1 | 5, 6 | 566 | |||
XM_011545356.2 | XP_011543658.1 | 6, 7 | 625 | |||
XM_011545357.2 | XP_011543659.1 | 6, 7 | 611 | |||
XM_011545358.2 | XP_011543660.1 | 6, 7 | 1149 | |||
XM_011545359.2 | XP_011543661.1 | 5, 6 | 1105 | |||
GenBank mRNA | AB207212.1 | - | 3, 4 | 321 | ||
AF087878.1 | - | 5, 6 | 596 | |||
AK055818.1 | - | 4, 5 | 718 | |||
AK091965.1 | - | 5, 6 | 464 | |||
AK130017.1 | - | 2, 3 | 1318 | |||
AK300479.1 | - | 6, 7 | 629 | |||
BC008353.1 | - | 6, 7 | 661 | 3537224 | ||
DQ084381.1 | - | 5, 6 | 561 |
Gene Symbol: GLYATL1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB231721.1 | - | 1, 2 | 81 |