Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145971.1 | NP_001139443.1 | 4 | 538 | ||
NM_138412.3 | NP_612421.1 | 5 | 630 | |||
NR_027381.1 | 4 | 538 | ||||
NR_027382.1 | 4 | 538 | ||||
XM_005258473.1 | XP_005258530.1 | 6 | 1019 | |||
XM_011526407.1 | XP_011524709.1 | 6 | 890 | |||
XM_011526408.2 | XP_011524710.1 | 4 | 1154 | |||
GenBank mRNA | AK075392.1 | - | 4 | 504 | ||
AK122764.1 | - | 4 | 1168 | |||
AK291579.1 | - | 4 | 421 | |||
AK310288.1 | - | 2 | 189 | |||
AK315539.1 | - | 2 | 163 | |||
AY358473.1 | - | 4 | 434 | |||
BC009881.2 | - | 5 | 614 | 3956119 |