Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127715.2 | NP_001121187.1 | 24 | 2853 | ||
NM_139244.4 | NP_640337.3 | 22 | 2745 | |||
XM_006715339.2 | XP_006715402.1 | 23 | 2979 | |||
XM_011535453.1 | XP_011533755.1 | 23 | 2577 | |||
XM_017010267.1 | XP_016865756.1 | 19 | 2924 | |||
GenBank mRNA | AB073398.1 | - | 13 | 1584 | ||
AK055484.1 | - | 5 | 563 | |||
AK090549.1 | - | 14 | 1563 | |||
AK128121.1 | - | 23 | 2572 | |||
AL834152.1 | - | 25 | 2902 | |||
BC113382.1 | - | 22 | 2618 | 8322434 | ||
BC113408.1 | - | 22 | 2618 | 8322460 |