Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323906.1 | NP_001310835.1 | 4, 5 | 836 | ||
NM_001323907.1 | NP_001310836.1 | 4, 5 | 716 | |||
NM_001323908.1 | NP_001310837.1 | 4, 5 | 868 | |||
NM_001323909.1 | NP_001310838.1 | 4, 5 | 836 | |||
NM_001323910.1 | NP_001310839.1 | 5, 6 | 884 | |||
NM_001323911.1 | NP_001310840.1 | 4, 5 | 836 | |||
NM_001323912.1 | NP_001310841.1 | 4, 5 | 787 | |||
NM_001323913.1 | NP_001310842.1 | 3, 4 | 668 | |||
NM_144618.2 | NP_653219.1 | 4, 5 | 810 | |||
XM_017000246.1 | XP_016855735.1 | 5, 6 | 1314 | |||
XM_017000247.1 | XP_016855736.1 | 6, 7 | 1401 | |||
XM_017000248.1 | XP_016855737.1 | 4, 5 | 710 | |||
XM_017000249.1 | XP_016855738.1 | 5, 6 | 770 | |||
XM_017000250.1 | XP_016855739.1 | 4, 5 | 527 | |||
XM_017000251.1 | XP_016855740.1 | 4, 5 | 710 | |||
XM_017000252.1 | XP_016855741.1 | 4, 5 | 556 | |||
XM_017000253.1 | XP_016855742.1 | 4, 5 | 527 | |||
XM_017000254.1 | XP_016855743.1 | 4, 5 | 649 | |||
GenBank mRNA | AK092230.1 | - | 4, 5 | 650 | ||
AK301882.1 | - | 5, 6 | 648 | |||
AK313606.1 | - | 4, 5 | 623 | |||
BC027033.1 | - | 4, 5 | 810 | 5139830 |