Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKAR SIRNA Assays ›
Gene Symbol:
ANKAR
Gene Name:
ankyrin and armadillo repeat containing
Gene Aliases:
-
Chromosome Location:
Chr.2: 189674292 - 189763059 on Build GRCh38
UniGene ID:
Hs.60772
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37537
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKAR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_144708.3
NP_653309.3
12
2692
XM_011510673.2
XP_011508975.1
12
3178
XM_011510674.1
XP_011508976.1
12
3178
XM_011510675.2
XP_011508977.1
11
3073
XM_011510676.2
XP_011508978.1
12
3178
XM_011510677.2
XP_011508979.1
12
3178
XM_011510679.2
XP_011508981.1
12
3178
XM_011510680.1
XP_011508982.1
12
3178
XM_011510681.1
XP_011508983.1
12
3178
XM_011510682.2
XP_011508984.1
12
3178
XM_011510685.2
XP_011508987.1
12
2734
XM_011510686.2
XP_011508988.1
12
3178
XM_011510687.2
XP_011508989.1
11
2542
XM_011510688.1
XP_011508990.1
12
3178
XM_011510689.2
XP_011508991.1
12
3178
XM_011510691.2
XP_011508993.1
8
1364
XM_011510692.2
XP_011508994.1
10
2378
XM_011510693.2
XP_011508995.1
9
2274
XM_011510694.2
XP_011508996.1
8
2093
XM_017003413.1
XP_016858902.1
12
3178
XM_017003414.1
XP_016858903.1
12
3178
XM_017003415.1
XP_016858904.1
7
1873
XM_017003416.1
XP_016858905.1
9
2285
XM_017003417.1
XP_016858906.1
12
3178
XM_017003418.1
XP_016858907.1
7
1988
XM_017003419.1
XP_016858908.1
7
1988
XM_017003420.1
XP_016858909.1
7
1891
GenBank mRNA
AJ549812.1
-
11
2391
AK058144.1
-
2
274
AK126073.1
-
12
2692
BC037215.1
-
9
2073
BC044907.1
-
11
2544
4836761
BC047413.1
-
13
1979
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-55874ff697-z77lg:80/10.239.161.145:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0