Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297608.1 | NP_001284537.1 | 12 | 2268 | ||
NM_145263.3 | NP_660306.1 | 13 | 2364 | |||
NR_123728.1 | 13 | 2414 | ||||
XM_005265716.1 | XP_005265773.1 | 11 | 2070 | |||
XM_005265717.2 | XP_005265774.1 | 9 | 1736 | |||
XM_005265718.1 | XP_005265775.1 | 9 | 1852 | |||
XM_011534369.1 | XP_011532671.1 | 12 | 2166 | |||
XM_011534370.1 | XP_011532672.1 | 13 | 2290 | |||
XM_011534372.1 | XP_011532674.1 | 10 | 1944 | |||
XM_017007731.1 | XP_016863220.1 | 11 | 1762 | |||
GenBank mRNA | AK304386.1 | - | 12 | 2183 | ||
BC025396.1 | - | 13 | 2271 | 4821894 | ||
BC037886.1 | - | 7 | 1043 |