Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143905.2 | NP_001137377.1 | 2, 3 | 455 | ||
NM_001194995.1 | NP_001181924.1 | 2, 3 | 748 | |||
NM_152269.4 | NP_689482.1 | 2, 3 | 539 | |||
XM_005253630.4 | XP_005253687.1 | 3, 4 | 559 | |||
XM_011538980.2 | XP_011537282.1 | 2, 3 | 1317 | |||
GenBank mRNA | AF061733.1 | - | 2, 3 | 381 | ||
AK095982.1 | - | 2, 3 | 919 | |||
AK126043.1 | - | 2, 3 | 4310 | |||
BC018145.1 | - | 2, 3 | 340 | 3846678 | ||
BC062329.1 | - | 2, 3 | 382 | 5484911 | ||
BI850864.1 | - | 2, 3 | 455 | |||
BU507560.1 | - | 2, 3 | 562 |