Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007018.4 | NP_008949.4 | 30 | 5064 | ||
XM_005251679.3 | XP_005251736.1 | 32 | 5336 | |||
XM_006716940.2 | XP_006717003.1 | 32 | 5336 | |||
XM_006716942.1 | XP_006717005.1 | 21 | 3565 | |||
XM_011518166.1 | XP_011516468.1 | 32 | 5336 | |||
XM_011518167.1 | XP_011516469.1 | 31 | 5177 | |||
XM_011518168.1 | XP_011516470.1 | 32 | 5261 | |||
XM_011518169.2 | XP_011516471.1 | 31 | 5211 | |||
XM_011518170.1 | XP_011516472.1 | 31 | 5159 | |||
XM_011518172.2 | XP_011516474.1 | 32 | 5336 | |||
XM_011518173.1 | XP_011516475.1 | 20 | 3411 | |||
XM_017014223.1 | XP_016869712.1 | 32 | 5336 | |||
XM_017014224.1 | XP_016869713.1 | 32 | 5336 | |||
XM_017014225.1 | XP_016869714.1 | 30 | 5022 | |||
XM_017014226.1 | XP_016869715.1 | 21 | 3565 | |||
XM_017014227.1 | XP_016869716.1 | 31 | 5354 | |||
XM_017014228.1 | XP_016869717.1 | 31 | 5210 | |||
GenBank mRNA | AF083322.1 | - | 7 | 1512 | ||
AF513978.1 | - | 30 | 5033 | |||
AK074079.1 | - | 21 | 3437 | |||
AK297330.1 | - | 1384 | ||||
BC137284.1 | - | 30 | 5057 | 9021281 | ||
BC137286.1 | - | 30 | 5056 | 9021283 | ||
BX640927.1 | - | 5 | 461 |