Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKMYT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ017689.1 | - | 1 | 149 |
Gene Symbol: PAQR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284511.1 | NP_001271440.1 | 3 | 2110 | ||
NM_001284512.1 | NP_001271441.1 | 2 | 2005 | |||
NM_001284513.1 | NP_001271442.1 | 3 | 1733 | |||
NM_001324118.1 | NP_001311047.1 | 4 | 2026 | |||
NM_152341.4 | NP_689554.2 | 3 | 2227 | |||
GenBank mRNA | AK054564.1 | - | 3 | 1811 | ||
AK092173.1 | - | 2 | 1692 | |||
AL832527.1 | - | 3 | 1840 | |||
BC033703.1 | - | 3 | 1885 | 4154019 |