Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152512.3 | NP_689725.2 | 6 | 1302 | ||
XM_006724149.3 | XP_006724212.1 | 6 | 1897 | |||
XM_006724150.3 | XP_006724213.1 | 6 | 1087 | |||
XM_006724151.1 | XP_006724214.1 | 5 | 791 | |||
XM_011529927.2 | XP_011528229.1 | 6 | 1396 | |||
XM_011529928.2 | XP_011528230.1 | 5 | 1153 | |||
XM_011529930.2 | XP_011528232.1 | 2 | 318 | |||
GenBank mRNA | AK093154.1 | - | 4 | 536 | ||
BC033895.2 | - | 6 | 1302 | 4831324 | ||
CR456452.1 | - | 5 | 1097 |