Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098514.2 | NP_001091984.2 | 3 | 698 | ||
NM_152459.4 | NP_689672.4 | 3 | 698 | |||
XM_005255143.3 | XP_005255200.2 | 3 | 524 | |||
XM_017022972.1 | XP_016878461.1 | 3 | 524 | |||
XM_017022973.1 | XP_016878462.1 | 3 | 524 | |||
XM_017022974.1 | XP_016878463.1 | 3 | 524 | |||
GenBank mRNA | AK075242.1 | - | 3 | 524 | ||
AK300708.1 | - | 3 | 472 | |||
AY358483.1 | - | 3 | 663 | |||
BC033681.1 | - | 3 | 593 | 5190553 |