Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152523.2 | NP_689736.1 | 5 | 453 | ||
XM_005246343.1 | XP_005246400.1 | 4 | 471 | |||
GenBank mRNA | AK097751.1 | - | 5 | 453 | ||
CN482952.1 | - | 4 | 593 |
Gene Symbol: LOC641367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024523.1 | 1 | 447 | |||
GenBank mRNA | BC024253.1 | - | 1 | 452 | ||
BC047098.1 | - | 1 | 408 |