Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139459.1 | NP_001132931.1 | 2 | 824 | ||
NM_152609.2 | NP_689822.2 | 2 | 698 | |||
XM_005273081.3 | XP_005273138.2 | 2 | 483 | |||
XM_005273083.4 | XP_005273140.1 | 2 | 607 | |||
XM_011544110.2 | XP_011542412.1 | 2 | 549 | |||
XM_011544111.1 | XP_011542413.1 | 2 | 720 | |||
XM_011544113.1 | XP_011542415.1 | 2 | 598 | |||
XM_017000487.1 | XP_016855976.1 | 2 | 594 | |||
XM_017000488.1 | XP_016855977.1 | 2 | 531 | |||
XM_017000489.1 | XP_016855978.1 | 2 | 598 | |||
GenBank mRNA | AK056563.1 | - | 2 | 531 | ||
AK074142.1 | - | 2 | 461 | |||
AL834246.2 | - | 2 | 532 | |||
BC010228.2 | - | 2 | 700 | 4151228 | ||
BC036200.1 | - | 2 | 591 | |||
BX647629.1 | - | 2 | 483 |