Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028407.2 | 10 | 2444 | |||
NR_045861.1 | 7 | 2335 | ||||
NR_045862.1 | 6 | 1469 | ||||
NR_045863.1 | 6 | 1463 | ||||
NR_045864.1 | 7 | 2377 | ||||
GenBank mRNA | AK125041.1 | - | 6 | 1267 | ||
AK126987.1 | - | 7 | 2198 | |||
BC032236.1 | - | 7 | 2114 | |||
BC036206.1 | - | 6 | 1465 | |||
CR933662.1 | - | 10 | 2419 |