Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153045.3 | NP_694590.2 | 9 | 1267 | ||
XM_006716997.1 | XP_006717060.1 | 9 | 1270 | |||
XM_011518350.1 | XP_011516652.1 | 10 | 1438 | |||
XM_011518351.2 | XP_011516653.1 | 10 | 1566 | |||
XM_011518352.1 | XP_011516654.1 | 10 | 1435 | |||
XM_011518353.1 | XP_011516655.1 | 9 | 1327 | |||
XM_011518354.1 | XP_011516656.1 | 9 | 1473 | |||
XM_011518355.1 | XP_011516657.1 | 9 | 1470 | |||
XM_017014429.1 | XP_016869918.1 | 10 | 1594 | |||
XM_017014430.1 | XP_016869919.1 | 8 | 1359 | |||
GenBank mRNA | AK095364.1 | - | 4 | 591 | ||
AK289383.1 | - | 9 | 1236 | |||
AL834412.1 | - | 7 | 879 | |||
BC019913.1 | - | 3 | 440 | |||
BC062550.1 | - | 5 | 609 | |||
BC107133.1 | - | 9 | 1205 | 40025594 | ||
BC107134.1 | - | 9 | 1418 | 40025600 | ||
BX649023.1 | - | 9 | 1153 |