Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BDNF SIRNA Assays ›
Gene Symbol:
BDNF
Gene Name:
brain derived neurotrophic factor
Gene Aliases:
ANON2, BULN2
Chromosome Location:
Chr.11: 27654893 - 27722058 on Build GRCh38
UniGene ID:
Hs.502182
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
40419
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BDNF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143805.1
NP_001137277.1
2
319
NM_001143806.1
NP_001137278.1
2
534
NM_001143807.1
NP_001137279.1
2
420
NM_001143808.1
NP_001137280.1
2
256
NM_001143809.1
NP_001137281.1
2
266
NM_001143810.1
NP_001137282.1
3
549
NM_001143811.1
NP_001137283.1
4
666
NM_001143812.1
NP_001137284.1
2
408
NM_001143813.1
NP_001137285.1
2
490
NM_001143814.1
NP_001137286.1
3
615
NM_001143816.1
NP_001137288.1
2
1013
NM_001709.4
NP_001700.2
2
508
NM_170731.4
NP_733927.1
2
830
NM_170732.4
NP_733928.1
2
617
NM_170733.3
NP_733929.1
2
520
NM_170734.3
NP_733930.1
2
468
NM_170735.5
NP_733931.1
1
1247
XM_011520280.2
XP_011518582.1
2
1414
GenBank mRNA
AF400438.1
-
1
180
AK289763.1
-
2
316
AK289853.1
-
2
217
AY054392.2
-
2
521
AY054393.2
-
2
263
AY054394.2
-
2
478
AY054395.2
-
2
561
AY054396.2
-
2
319
AY054397.2
-
2
402
AY054398.2
-
2
449
AY054399.2
-
3
334
AY054400.2
-
2
435
AY054406.3
-
2
303
AY656701.1
-
1
162
BC029795.1
-
2
493
5193877
CX751420.1
-
2
387
EF674517.1
-
2
319
EF674518.1
-
2
534
EF674519.1
-
2
617
EF674520.1
-
2
420
EF674521.1
-
2
520
EF689009.1
-
1
1261
EF689010.1
-
2
1027
EF689011.1
-
2
256
EF689012.1
-
2
266
EF689013.1
-
2
408
EF689014.1
-
2
490
EF689015.1
-
2
508
EF689016.1
-
3
615
EF689017.1
-
3
367
EF689018.1
-
2
468
EF689019.1
-
3
549
EF689020.1
-
4
666
EF689021.1
-
2
830
KP229412.1
-
1
162
KU177943.1
-
1
162
KU177944.1
-
1
162
KU177945.1
-
1
162
M61176.1
-
1
724
X60201.1
-
2
458
X91251.1
-
2
446
Back To Top
More Information
Panther Classification:
Molecular Function -
neurotrophic factor
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell signaling
nervous system development
axon guidance
synapse assembly
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of brain-derived neurotrophic factor receptor signaling pathway
negative regulation of neuron apoptotic process
collateral sprouting
positive regulation of collateral sprouting
positive regulation of synapse assembly
neurotrophin TRKB receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0