Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199777.1 | NP_001186706.1 | 8 | 1278 | ||
NM_172240.2 | NP_758440.1 | 9 | 1297 | |||
NR_037659.1 | 8 | 1125 | ||||
NR_037660.1 | 7 | 1106 | ||||
GenBank mRNA | AB053266.1 | - | 8 | 787 | ||
AK027829.1 | - | 5 | 496 | |||
AK074772.1 | - | 9 | 1035 | |||
AK123106.1 | - | 8 | 1275 | |||
AK123625.1 | - | 5 | 495 | |||
AK295596.1 | - | 8 | 863 | |||
AL832918.1 | - | 7 | 762 | |||
BC026080.2 | - | 9 | 1059 | 4827689 | ||
EF011619.1 | - | 9 | 1059 |