Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289002.1 | NP_001275931.1 | 2 | 585 | ||
NM_001289004.1 | NP_001275933.1 | 2 | 585 | |||
NM_001308014.1 | NP_001294943.1 | 2 | 585 | |||
NM_173488.4 | NP_775759.3 | 2 | 585 | |||
XM_005271874.3 | XP_005271931.1 | 2 | 3782 | |||
XM_011543147.2 | XP_011541449.1 | 2 | 3782 | |||
XM_011543148.2 | XP_011541450.1 | 2 | 3782 | |||
XM_011543154.2 | XP_011541456.1 | 2 | 3782 | |||
GenBank mRNA | AF505657.1 | - | 2 | 556 | ||
AK131445.1 | - | 2 | 482 | |||
AY273897.1 | - | 2 | 548 | |||
BC034976.2 | - | 2 | 587 | 4820512 | ||
BM554439.1 | - | 2 | 572 |