Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK32C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318878.1 | NP_001305807.1 | 12 | 2085 | ||
NM_001318879.1 | NP_001305808.1 | 12 | 1946 | |||
NM_173575.3 | NP_775846.2 | 12 | 2047 | |||
NR_134911.1 | 13 | 1794 | ||||
XM_011539688.1 | XP_011537990.1 | 12 | 2118 | |||
XM_011539690.2 | XP_011537992.1 | 12 | 2033 | |||
XM_011539693.2 | XP_011537995.1 | 12 | 2051 | |||
XM_011539694.1 | XP_011537996.1 | 12 | 1785 | |||
XM_011539695.1 | XP_011537997.1 | 12 | 1986 | |||
XM_011539696.1 | XP_011537998.1 | 12 | 1793 | |||
GenBank mRNA | AK057849.1 | - | 13 | 1794 | ||
AK098366.1 | - | 10 | 2074 | |||
AY098866.1 | - | 12 | 2047 | |||
BC045760.1 | - | 12 | 1936 | 4831007 |