Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317056.1 | NP_001303985.1 | 1 | 220 | ||
NR_133652.1 | 1 | 220 | ||||
XM_011516065.1 | XP_011514367.1 | 1 | 220 | |||
XM_011516066.2 | XP_011514368.1 | 1 | 220 | |||
GenBank mRNA | AK096734.1 | - | 1 | 220 | ||
AY515311.1 | - | 1 | 220 |