Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000605.3 | NP_000596.2 | 1 | 806 | ||
GenBank mRNA | BC104163.1 | - | 1 | 783 | 40029595 | |
BC104164.1 | - | 1 | 783 | 40029596 | ||
M54886.1 | - | 1 | 799 | |||
V00544.1 | - | 1 | 799 | |||
V00548.1 | - | 1 | 720 | |||
V00549.1 | - | 1 | 798 |