Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173667.3 | NP_775938.1 | 4 | 408 | ||
NR_126523.1 | 3 | 220 | ||||
NR_126524.1 | 3 | 220 | ||||
XM_017009378.1 | XP_016864867.1 | 2 | 2827 | |||
XM_017009379.1 | XP_016864868.1 | 2 | 2827 | |||
XM_017009380.1 | XP_016864869.1 | 2 | 2827 | |||
XM_017009381.1 | XP_016864870.1 | 2 | 2827 | |||
XM_017009382.1 | XP_016864871.1 | 3 | 329 | |||
GenBank mRNA | AK094862.1 | - | 4 | 380 | ||
BC111982.1 | - | 3 | 334 | 8327450 | ||
DA152848.1 | - | 3 | 221 | |||
DB463100.2 | - | 3 | 192 |