Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACKR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001296.4 | NP_001287.2 | 3 | 1258 | ||
GenBank mRNA | AK313561.1 | - | 3 | 1255 | ||
BC008816.2 | - | 3 | 1246 | 3959017 | ||
BC011631.2 | - | 3 | 1246 | 3958671 | ||
BC020558.1 | - | 2 | 1177 | 3451472 | ||
BC101629.1 | - | 1 | 1110 | 8069135 | ||
BC112045.1 | - | 1 | 1110 | 8327513 | ||
BT006800.1 | - | 1 | 1079 | |||
U94888.1 | - | 1 | 1094 | |||
Y12815.1 | - | 1 | 1091 |